Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGRP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB036570 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036570 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036570 Supplier Novus Biologicals Supplier No. NBP288979
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CGRP1 Polyclonal antibody specifically detects CGRP1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen CGRP1
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Antiserum
Gene Alias alpha-type CGRP, CALCA, calcitonin gene-related peptide I, calcitonin/calcitonin-related polypeptide, alpha, calcitonin; calcitonin gene-related peptide 1, CGRP-I
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a portion of rat alpha-calcitonin gene-related peptide (CGRP).
Purification Method Whole Antiserum
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cellular Signaling, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 796
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.