Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ch 55
GREENER_CHOICE

Catalog No. 202010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-201-0 10 mg
20-205-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 20-201-0 Supplier Tocris Bioscience™ Supplier No. 2020/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent RAR agonist

Highly potent synthetic retinoid that has high affinity for RAR-α and RAR-β receptors and low affinity for cellular retinoic acid binding protein (CRABP). Inhibits rabbit tracheal epithelial cell differentiation by inhibiting transglutaminase and increasing cholesterol sulfate (EC50 values are 0.02 and 0.03 nM respectively). Induces differentiation of embryonic carcinoma F9 and melanoma S91 cells (EC50 values are 0.26 and 0.5 nM respectively) and inhibits the induction of ornithine decarboxylase activity in 3T6 fibroblasts (EC50 = 1 nM).

Chemical Identifiers

CAS 110368-33-7
Molecular Formula C24H28O3

Specifications

Quantity 10 mg
Formula Weight 364.47
Purity >98%
Molecular Weight (g/mol) 364.485
Chemical Name or Material Ch 55
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.