Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ch-TOG Antibody, Novus Biologicals™
SDP

Catalog No. NB500182 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500182 0.1 mL
NB500182SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500182 Supplier Novus Biologicals Supplier No. NB500182
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

ch-TOG Polyclonal specifically detects ch-TOG in Human, Mouse, Rat, Mammal samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen ch-TOG
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1:10-1:500
Formulation Whole antisera with No Preservative
Gene Accession No. Q14008
Gene Alias Ch-TOG, CHTOG, Colonic and hepatic tumor over-expressed gene protein, cytoskeleton associated protein 5, cytoskeleton-associated protein 5, EC 2.7.7.6, EC 3.1.1.29, KIAA0097FLJ35359, MSPS, TOG, TOGp
Gene Symbols CKAP5
Host Species Rabbit
Immunogen A recombinant segment of the C-terminal domain of human ch-TOG [UniProt# Q14008]
Molecular Weight of Antigen 200 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9793
Target Species Human, Mouse, Rat, Mammalia
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.