Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CH25H Antibody, Novus Biologicals™
SDP

Catalog No. NB154866 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB154866 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB154866 Supplier Novus Biologicals Supplier No. NBP305283
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CH25H Polyclonal antibody specifically detects CH25H in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen CH25H
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:10000-1:50000, Immunohistochemistry 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Gene Alias C25H, cholesterol 25-hydroxylase, Cholesterol 25-monooxygenase, EC 1.14.99.38, h25OH
Host Species Rabbit
Immunogen CH25H Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a C-Terminal region of human CH25H conjugated to Keyhole Limpet Hemocyanin (KLH). (Uniprot: O95992)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9023
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.