Learn More
CH25H Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $661.74
Specifications
| Antigen | CH25H |
|---|---|
| Concentration | 0.92 mg/mL |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2328
![]()
|
Rockland Immunochemicals
600401MM8 |
100 μg |
Each for $661.74
|
|
|||||
50-199-2329
![]()
|
Rockland Immunochemicals
600401MM8S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human CH25H. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence and 92% homology to pig, 91% homology to rat, and 75% homology to mouse.
Catalyzes the formation of 25-hydroxycholesterol from cholesterol, leading to repress cholesterol biosynthetic enzymes (PubMed:9852097). Plays a key role in cell positioning and movement in lymphoid tissues: 25-hydroxycholesterol is an intermediate in biosynthesis of 7-alpha,25-dihydroxycholesterol (7-alpha,25-OHC), an oxysterol that acts as a ligand for the G protein-coupled receptor GPR183/EBI2, a chemotactic receptor for a number of lymphoid cells. May play an important role in regulating lipid metabolism by synthesizing a corepressor that blocks sterol regulatory element binding protein (SREBP) processing. In testis, production of 25-hydroxycholesterol by macrophages may play a role in Leydig cell differentiation.Specifications
| CH25H | |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O95992 | |
| 9023 | |
| Primary | |
| Antibody |
| 0.92 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| 1.14.99.38; AI462618; BOS_23282; C25H; ch25h; ch25hb; Cholesterol 25-hydroxylase; cholesterol 25-hydroxylase-like; Cholesterol 25-hydroxylase-like protein; Cholesterol 25-monooxygenase; h25OH; LOC100341039; m25OH; zgc:101688 | |
| CH25H | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| CH25H |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.