Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChABC Antibody (6A12), FITC, Novus Biologicals™
SDP

Catalog No. NBP196142F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP196142F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP196142F Supplier Novus Biologicals Supplier No. NBP196142F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ChABC Monoclonal specifically detects ChABC in Non-species specific samples. It is validated for ELISA.

Specifications

Antigen ChABC
Applications ELISA
Classification Monoclonal
Clone 6A12
Conjugate FITC
Dilution ELISA
Gene Alias Chondroitinase ABC
Host Species Mouse
Immunogen Chondroitinase ABC
Molecular Weight of Antigen 80 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.