Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChABC Antibody (6A12), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB337756 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337756 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337756 Supplier Novus Biologicals Supplier No. NBP196142MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ChABC Monoclonal antibody specifically detects ChABC in Nonspecies specific samples. It is validated for Western Blot, ELISA

Specifications

Antigen ChABC
Applications Western Blot, ELISA
Classification Monoclonal
Clone 6A12
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Chondroitinase ABC
Host Species Mouse
Immunogen Chondroitinase ABC
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.