Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHAF1A Antibody, Novus Biologicals™
SDP

Catalog No. NB10074608 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074608 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074608 Supplier Novus Biologicals Supplier No. NB10074608
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHAF1A Polyclonal specifically detects CHAF1A in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen CHAF1A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:200 - 1:1000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:200 - 1:1000
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. AAA76736.1
Gene Alias CAF, CAF1, CAF-1, CAF-1 subunit A, CAF1B, CAF1P150CAF-I 150 kDa subunit, CAF-I p150, chromatin assembly factor 1 subunit A, chromatin assembly factor 1, subunit A (p150), chromatin assembly factor I (150 kDa), Chromatin assembly factor I p150 subunit, hp150, MGC71229, P150
Gene Symbols CHAF1A
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human chromatin assembly factor 1, subunit A (p150) using the numbering given in entry AAA76736.1 (GeneID 10036).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10036
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.