Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHAF1A Antibody (SS 1 1-13), Novus Biologicals™
SDP

Catalog No. NB500207SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500207SS 0.05 mL
NB500207 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500207SS Supplier Novus Biologicals Supplier No. NB500207SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 9 publications

CHAF1A Monoclonal specifically detects CHAF1A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen CHAF1A
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SS 1 1-13
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. Q13111
Gene Alias CAF, CAF1, CAF-1, CAF-1 subunit A, CAF1B, CAF1P150CAF-I 150 kDa subunit, CAF-I p150, chromatin assembly factor 1 subunit A, chromatin assembly factor 1, subunit A (p150), chromatin assembly factor I (150 kDa), Chromatin assembly factor I p150 subunit, hp150, MGC71229, P150
Gene Symbols CHAF1A
Host Species Mouse
Immunogen Recombinant human CHAF1A [UniProt# Q13111]
Molecular Weight of Antigen 150 kDa
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10036
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.