Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHAF1B Antibody (OTI4F7), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB27136A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB27136A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB27136A700 Supplier Novus Biologicals Supplier No. NBP271366AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CHAF1B Monoclonal specifically detects CHAF1B in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CHAF1B
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4F7
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CAF1, CAF-1, CAF1ACAF-1 subunit B, CAF1P60CAF-I 60 kDa subunit, CAF-IP60, chromatin assembly factor 1 subunit B, chromatin assembly factor 1, subunit B (p60), Chromatin assembly factor I p60 subunit, human chromatin assembly factor-I p60 subunit, 10, MPHOSPH7CAF-I p60, MPP7M-phase phosphoprotein 7
Gene Symbols CHAF1B
Host Species Mouse
Immunogen Full length recombinant protein of human CHAF1B (NP_005432) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 8208.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.