Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHAF1B Antibody (SS 53), Novus Biologicals™
SDP

Catalog No. NB500212 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500212 0.1 mL
NB500212SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500212 Supplier Novus Biologicals Supplier No. NB500212
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 8 publications

CHAF1B Monoclonal specifically detects CHAF1B in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockdown Validated.

Specifications

Antigen CHAF1B
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SS 53
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 1:10-1:500, KnockDown Validated
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. Q13112
Gene Alias CAF1, CAF-1, CAF1ACAF-1 subunit B, CAF1P60CAF-I 60 kDa subunit, CAF-IP60, chromatin assembly factor 1 subunit B, chromatin assembly factor 1, subunit B (p60), Chromatin assembly factor I p60 subunit, human chromatin assembly factor-I p60 subunit, 10, MPHOSPH7CAF-I p60, MPP7M-phase phosphoprotein 7
Gene Symbols CHAF1B
Host Species Mouse
Immunogen Recombinant human CHAF1B [UniProt# Q13112]
Molecular Weight of Antigen 60 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 8208
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.