Learn More
Description
Specifications
Specifications
| Antigen | CHAF1B |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | SS 53 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 1:10-1:500, KnockDown Validated |
| Formulation | Ascites with 0.1% Sodium Azide |
| Gene Accession No. | Q13112 |
| Gene Alias | CAF1, CAF-1, CAF1ACAF-1 subunit B, CAF1P60CAF-I 60 kDa subunit, CAF-IP60, chromatin assembly factor 1 subunit B, chromatin assembly factor 1, subunit B (p60), Chromatin assembly factor I p60 subunit, human chromatin assembly factor-I p60 subunit, 10, MPHOSPH7CAF-I p60, MPP7M-phase phosphoprotein 7 |
| Gene Symbols | CHAF1B |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
