Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Changes in State STEM Kit
SDPSCI_ED

Catalog No. S43774 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
8 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S43774 8 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43774 Supplier United Scientific Supplies Supplier No. AISCSKIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains two investigative areas and six STEM investigations that teach students about heats of fusion and reaction, thermochemistry, iceberg types and thermohaline circulation and climate change.

  • Contains two investigative areas and six STEM investigations that teach students about heats of fusion and reaction, thermochemistry, iceberg types and thermohaline circulation and climate change.
  • The kit includes enough materials for 40 students working in groups of four, as well as PDF teacher and student guides and other digital content.
  • Skills/Concepts: Experimental/Engineering Design, Investigating, Physical State, Scientific Method, Measuring, Data Analysis, Spreadsheet Preparation, Communication, Technology, Developing & Using Models, Using a Calorimeter.
  • Activity 1- Learning About Heat of Fusion
  • Activity 2- Climate Change: Modeling Icebergs Melting in Seawater
  • Activity 3- Is Ice Melting an Exo or Endothermic Reaction?
  • Activity 4- Determine The Most Efficient Ice-Melt Product
  • Activity 5- (STEM Activity)- Designing an Ice-Melt Strategy for City Sidewalks
  • Activity 6- The Eutectics of Ice Cream

Specifications

Grade 8 to 12
Format Kit
Product Type Changes In State Set
Languages English
Subject STEM, Physics - Matter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.