Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHAPS, 98+%

Catalog No. AAB2192706
Click to view available options
Quantity:
1 g
5 g
25 g

CHAPS, >-98%, C32H58N2O7S, CAS Number-75621-03-3, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, 25g, 614.88, H315-H319-H335, GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. | CAS: 75621-03-3 | C32H58N2O7S | 614.883 g/mol

Buffer; zwitterionic nondenaturing detergent for solubilizing membrane proteinsCHAPS is used in the laboratory to dissolve biological macromolecules such as proteins. It acts as a non-denaturing solvent used in purification of membrane proteins. It is also useful in conjunction with nonionic detergents like Triton X-100. Further, it is used as a non-denaturing zwitterionic detergent for membrane biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Buffer; zwitterionic nondenaturing detergent for solubilizing membrane proteinsCHAPS is used in the laboratory to dissolve biological macromolecules such as proteins. It acts as a non-denaturing solvent used in purification of membrane proteins. It is also useful in conjunction with nonionic detergents like Triton X-100. Further, it is used as a non-denaturing zwitterionic detergent for membrane biochemistry.

Solubility
Slightly soluble in water.

Notes
Store in a cool. Incompatible with oxidizing agent, strong acid and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75621-03-3
Molecular Formula C32H58N2O7S
Molecular Weight (g/mol) 614.883
MDL Number MFCD00012116
InChI Key UMCMPZBLKLEWAF-GBSSQDIOSA-N
Synonym 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
PubChem CID 134129639
IUPAC Name 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate
SMILES CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Melting Point 156°C to 158°C (decomposition)
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 614.88
Percent Purity ≥98%
Chemical Name or Material CHAPS
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.