Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CHC
GREENER_CHOICE

Catalog No. 502950r
Click to view available options
:
50mg
This item is not returnable. View return policy
This item is not returnable. View return policy

MCT inhibitor; decreases glycoloysis

CHC is a monocarboxylic acid transport (MCT) inhibitor. Exhibits antitumoral and antiangiogenic activity in gliomas in vivo; decreases glycolytic metabolism, migration, and invasion in U251 cells. Enhances the effect of antitumor agent temozolomide (Cat No. 2706). Blocks lactate efflux from glioma cells and sensitizes cells to irradiation.

Specifications

Quantity 50 mg
Product Type CHC
Purity >98%
Molecular Weight (g/mol) 189.17
Inhibitors CHC

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.