Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHCHD3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152008 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152008 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152008 Supplier Novus Biologicals Supplier No. NBP152008
Add to Cart
Add to Cart

Goat Polyclonal Antibody

CHCHD3 Polyclonal specifically detects CHCHD3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA, Immunohistochemistry-Frozen (Negative).

Specifications

Antigen CHCHD3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/mL, Immunohistochemistry 5 ug/mL, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:16000, Immunohistochemistry-Frozen (Negative) 5 ug/mL
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_060282.1
Gene Alias coiled-coil-helix-coiled-coil-helix domain containing 3, FLJ20420, mitochondrial
Gene Symbols CHCHD3
Host Species Goat
Immunogen Peptide with sequence KFKRYESHPVCAD corresponding to internal region according to NP_060282.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 54927
Test Specificity Cross reacts with Human. Expected from sequence similarity: Rat, Dog, Cow and Sheep.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.