Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHCHD3 Antibody (OTI7G4), FITC, Novus Biologicals™
SDP

Catalog No. NB288490 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288490 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288490 Supplier Novus Biologicals Supplier No. NBP271975F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CHCHD3 Monoclonal antibody specifically detects CHCHD3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen CHCHD3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7G4
Conjugate FITC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias coiled-coil-helix-coiled-coil-helix domain containing 3, FLJ20420, mitochondrial
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 2-227 of human CHCHD3 (NP_060282) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 54927
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.