Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHCHD5 Antibody (OTI3H3), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N272289APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272289APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272289APC7 Supplier Novus Biologicals Supplier No. NBP272289APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CHCHD5 Monoclonal specifically detects CHCHD5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen CHCHD5
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3H3
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias C2orf9, chromosome 2 open reading frame 9, coiled-coil-helix-coiled-coil-helix domain containing 5, coiled-coil-helix-coiled-coil-helix domain-containing protein 5, FLJ39671, MGC11104
Gene Symbols CHCHD5
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-110 of human CHCHD5 (NP_115685) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84269.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.