Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHCHD6 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB341640 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341640 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341640 Supplier Novus Biologicals Supplier No. NBP281977PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHCHD6 Polyclonal antibody specifically detects CHCHD6 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen CHCHD6
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias coiled-coil-helix-coiled-coil-helix domain containing 6, coiled-coil-helix-coiled-coil-helix domain-containing protein 6, MGC13016
Host Species Rabbit
Immunogen Rabbit polyclonal CHCHD6 antibody was raised against a 17 amino acid peptide near the amino terminus of human CHCHD6. The immunogen is located within amino acids 20 - 70 of CHCHD6. Amino Acid Squence: VNRMKEPSSPPAPTSST
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 84303
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.