Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10060411 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10060411 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10060411 Supplier Novus Biologicals Supplier No. NB10060411
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

CHD1 Polyclonal specifically detects CHD1 in Human, Mouse samples. It is validated for Chemotaxis, ChIP Assay, ChIP sequencing (ChIP-seq), Gene Knock-Out, Immunoprecipitation, KnockDown, Western Blot.

Specifications

Antigen CHD1
Applications Chemotaxis, ChIP Assay, ChIP sequencing (ChIP-seq), Gene Knock-Out, Immunoprecipitation, KnockDown, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate, Chromatin Immunoprecipitation (ChIP) 1-5 ug/mg lysate, Knockout Validated, Chromatin Immunoprecipitation Sequencing, Knockdown Validated
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. O14646
Gene Alias ATP-dependent helicase CHD1, CHD-1, chromodomain helicase DNA binding protein 1, chromodomain-helicase-DNA-binding protein 1, DKFZp686E2337, EC 3.6.1, EC 3.6.4.12
Gene Symbols CHD1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1660 and 1710 of human chromodomain helicase DNA binding protein 1 using the numbering given in entry NP_001261.2 (GeneID 1105).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1105
Test Specificity Mouse reactivity reported in scientific literature (PMID: 20161795). Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Gorilla, Tasmanian devil, White-tufted-ear marmoset, Crab-eating macaque, African elephant and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.