Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD1L Rabbit anti-Human, Mouse, Clone: 3C1O9, Novus Biologicals™
SDP

Catalog No. NB166177 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166177 100 μg
NB166178 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166177 Supplier Novus Biologicals Supplier No. NBP315541100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CHD1L Monoclonal antibody specifically detects CHD1L in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen CHD1L
Applications Western Blot
Classification Monoclonal
Clone 3C1O9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ALC1, amplified in liver cancer 1, amplified in liver cancer protein 1, CHDL, chromodomain helicase DNA binding protein 1-like, chromodomain-helicase-DNA-binding protein 1-like, FLJ22530
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 700-800 of human CHD1L (Q86WJ1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9557
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.