Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHD1L Rabbit anti-Human, Mouse, Clone: 3C1O9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | CHD1L |
---|---|
Clone | 3C1O9 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CHD1L Monoclonal antibody specifically detects CHD1L in Human, Mouse samples. It is validated for Western BlotSpecifications
CHD1L | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
ALC1, amplified in liver cancer 1, amplified in liver cancer protein 1, CHDL, chromodomain helicase DNA binding protein 1-like, chromodomain-helicase-DNA-binding protein 1-like, FLJ22530 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 700-800 of human CHD1L (Q86WJ1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
3C1O9 | |
Monoclonal | |
Purified | |
RUO | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
9557 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title