Learn More
Description
Specifications
Specifications
| Antigen | CHD2 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | ATP-dependent helicase CHD2, CHD-2, chromodomain helicase DNA binding protein 2, chromodomain-helicase-DNA-binding protein 2, DKFZp547I1315, DKFZp686E01200, DKFZp781D1727, EC 3.6.1, EC 3.6.4.12, FLJ38614 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 242-501 of human CHD2 (NP_001036037.1). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
