Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHD3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CHD3 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CHD3 Polyclonal specifically detects CHD3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
CHD3 | |
Polyclonal | |
Rabbit | |
Immune Dysfunction, Immune System Diseases, Immunology | |
ATP-dependent helicase CHD3, CHD-3, chromodomain helicase DNA binding protein 3, chromodomain-helicase-DNA-binding protein 3, EC 3.6.1, EC 3.6.4.12, hZFH, Mi-2 autoantigen 240 kDa protein, Mi-2a, Mi2-ALPHA, ZFH, Zinc finger helicase, zinc-finger helicase (Snf2-like) | |
CHD3 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
1107 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:('PAPSEKGEGIRTPLEKEEAENQEEKPEKNSRIGEKMETEADAPSPAPSLGERLEPRKIPLEDEVPGVPGEMEPEPGYRGDREKSATESTPGERGEEKPLDG',) | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title