Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD3 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291677 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291677 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291677 Supplier Novus Biologicals Supplier No. NB10060412PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHD3 Polyclonal antibody specifically detects CHD3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen CHD3
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ATP-dependent helicase CHD3, CHD-3, chromodomain helicase DNA binding protein 3, chromodomain-helicase-DNA-binding protein 3, EC 3.6.1, EC 3.6.4.12, hZFH, Mi-2 autoantigen 240 kDa protein, Mi-2a, Mi2-ALPHA, ZFH, Zinc finger helicase, zinc-finger helicase (Snf2-like)
Host Species Rabbit
Immunogen Maps to a region between residue 1575 and 1625 of human chromodomain helicase DNA binding protein 3 using the numbering given in entry NP_001005273.1 (GeneID 1107).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune Dysfunction, Immune System Diseases, Immunology, modENCODE Antibodies
Primary or Secondary Primary
Gene ID (Entrez) 1107
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.