Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD3 Rabbit anti-Human, Mouse, Rat, Clone: 8Z6L6, Novus Biologicals™
SDP

Catalog No. NB168355 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168355 100 μg
NB168356 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168355 Supplier Novus Biologicals Supplier No. NBP316632100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CHD3 Monoclonal antibody specifically detects CHD3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CHD3
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8Z6L6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ATP-dependent helicase CHD3, CHD-3, chromodomain helicase DNA binding protein 3, chromodomain-helicase-DNA-binding protein 3, EC 3.6.1, EC 3.6.4.12, hZFH, Mi-2 autoantigen 240 kDa protein, Mi-2a, Mi2-ALPHA, ZFH, Zinc finger helicase, zinc-finger helicase (Snf2-like)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CHD3 (Q12873).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immune Dysfunction, Immune System Diseases, Immunology, modENCODE Antibodies
Primary or Secondary Primary
Gene ID (Entrez) 1107
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.