Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD4 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB327079 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327079 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327079 Supplier Novus Biologicals Supplier No. NBP299099MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHD4 Polyclonal antibody specifically detects CHD4 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen CHD4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ATP-dependent helicase CHD4, CHD-4, chromodomain helicase DNA binding protein 4, chromodomain-helicase-DNA-binding protein 4, DKFZp686E06161, EC 3.6.1, EC 3.6.4.12, Mi-2 autoantigen 218 kDa protein, Mi-2b, Mi2-BETA
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CHD4.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1108
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.