Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chd5 Monoclonal Antibody (5A10), Active Motif™
SDP

Rat Monoclonal Antibody

$156.65 - $620.58

Specifications

Antigen Chd5
Clone 5A10
Concentration 1 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3617
SDP
Encompass
View Documents
Active Motif
61684
10 μg
Each for $156.65
Only null left
Add to Cart
 
50-199-3616
SDP
Encompass
View Documents
Active Motif
61683
100 μg
Each for $620.58
Only null left
Add to Cart
 
Description

Description

CHD5 (Chromodomain helicase DNA binding protein 5) is a member of the SNF2/RAD54 helicase family of chromatin remodeling and DNA-binding proteins (CDH proteins). Heavily expressed in both fetal and adult brain, CHD5 plays a role in nervous system development and acts as a tumor suppressor via the Arf/p53 pathway. CHD5, along with other chromo-domain proteins, forms remodeling complexes, such as NuRD, that promote normal neuroblast maturation and are thought to prevent overexpression of neuronal cells. Errors in these chromatin remodeling complexes can leave the cell in a perpetual state of growth, preventing differentiation and leading to tumor formation. Due to the importance of the CHD proteins in proper brain development, deletions in the gene encoding CHD5 are commonly found in neuroblastomas, suggesting that CHD5 deficiency may lead to malignant cell transformation and metastasis.
Specifications

Specifications

Chd5
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
4930532L22Rik; ATP-dependent helicase CHD5; AW060752; B230399N07Rik; CHD5; CHD-5; chromodomain helicase DNA binding protein 5; Chromodomain-helicase-DNA-binding protein 5; KIAA0444
CHD5
IgG2a
Protein G
Antibody
5A10
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rat
Human, Mouse
A2A8L1, Q8TDI0
26038, 269610
This antibody was raised against a peptide within the C-terminal region of mouse Chd5.
Primary
-20°C, Avoid Freeze/Thaw Cycles
CHD5
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.