Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD7 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP177393R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177393R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177393R Supplier Novus Biologicals Supplier No. NBP177393R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHD7 Polyclonal specifically detects CHD7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CHD7
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9P2D1
Gene Alias ATP-dependent helicase CHD7, CHD-7, chromodomain helicase DNA binding protein 7, chromodomain helicase DNA binding protein 7 isoform CRA_e, chromodomain-helicase-DNA-binding protein 7, EC 3.6.1, EC 3.6.4.12, FLJ20357, FLJ20361, IS3, KIAA1416KAL5
Gene Symbols CHD7
Host Species Rabbit
Immunogen A genomic peptide made to an N-terminal region of the human Chd7 protein (within residues 25-200). [Swiss-Prot Q9P2D1]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 55636
Test Specificity Human and mouse. Immunogen has 85% identity to chicken Chd7.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.