Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD8 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB10060417F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10060417F 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10060417F Supplier Novus Biologicals Supplier No. NB10060417F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHD8 Polyclonal specifically detects CHD8 in Human, Mouse samples. It is validated for ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry.

Specifications

Antigen CHD8
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias ATP-dependent helicase CHD8, CHD-8, chromodomain helicase DNA binding protein 8, DKFZp686N17164, duplin, EC 3.6.1, EC 3.6.1.7, EC 3.6.4.12, Helicase with SNF2 domain 1chromodomain-helicase-DNA-binding protein 8, HELSNF1, KIAA1564axis duplication inhibitor
Gene Symbols CHD8
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 325 and 350 of human chromodomain helicase DNA binding protein 8 (NP_065971.2).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 57680
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.