Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemerin Antibody (13G2), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB221989647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB221989647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB221989647 Supplier Novus Biologicals Supplier No. NBP221989AF647
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Chemerin Monoclonal specifically detects Chemerin in Mouse samples. It is validated for ELISA.

Specifications

Antigen Chemerin
Applications ELISA
Classification Monoclonal
Clone 13G2
Conjugate Alexa Fluor 647
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias chemerin, RAR-responsive protein TIG2, retinoic acid receptor responder (tazarotene induced) 2, retinoic acid receptor responder protein 2, Tazarotene-induced gene 2 protein, TIG2HP10433
Gene Symbols RARRES2
Host Species Rat
Immunogen Mouse recombinant Chemerin
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cardiovascular Biology, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5919
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.