Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ Chemical Hazard Identification Labels

Catalog No. 1798159E Shop All National Marker Company Products
Click to view available options
Product Type:
Acetone Container Label
Bleach solid Container Label
Diesel fuel Container Label
Ethyl alcohol Container Label
Gasoline Container Label
Hydrochloric acid Container Label
Isopropyl alcohol Container Label
Kerosene Container Label
Methyl ethyl ketone Container Label
Sulfuric acid Container Label

High-visibility red-and-white pressure-sensitive vinyl labels ideal for alerting employees to potential chemical hazards in the workplace

  • Withstand most chemicals, solvents and vapors
  • Resist fading and scratching
  • For medium-sized containers
  • Measure 3 × 5 in.

Specifications

Color White, Red
Format Label
Material Vinyl
Product Type Ethyl alcohol Container Label
For Use With (Application) Hazard identification, suitable for both indoor and outdoor applications
Quantity 10/Pk.
Thickness (English) 0.0045 mil
Width (English) 5 in.
Height (English) 3 in.
Height (Metric) 7.62 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.