Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemR23/CMKLR1 Antibody (BZ194), Biotin, Novus Biologicals™
SDP

Catalog No. NB338401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338401 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338401 Supplier Novus Biologicals Supplier No. NBP143233B
Add to Cart
Add to Cart

Rat Monoclonal Antibody

ChemR23/CMKLR1 Monoclonal antibody specifically detects ChemR23/CMKLR1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ChemR23/CMKLR1
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BZ194
Conjugate Biotin
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, DEZ, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez
Host Species Rat
Immunogen Partial synthetic peptide made to an N-terminal portion of the mouse ChemR23/CMKLR1 protein (amino acids 8-32) [UniProt P97468]
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1240
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.