Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals CHES, 0.5M buffer soln, pH 9.0
Click to view available options
Quantity:
100 mL
250 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Concentration | 0.5 M |
| pH | 9.0 |
| Physical Form | Liquid |
| Quantity | 250 mL |
| Chemical Name or Material | CHES buffer soln. |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement P280-P264-P305+P351+P338-P337+P313 Wear protective gloves/protective clothing/eye protection/face protection. Wash thoroughly after handling. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction