Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHFR Antibody, Novus Biologicals™
SDP

Catalog No. NB392808 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392808 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392808 Supplier Novus Biologicals Supplier No. NBP26897025UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHFR Polyclonal antibody specifically detects CHFR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen CHFR
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias checkpoint with forkhead and ring finger domains, Checkpoint with forkhead and RING finger domains protein, E3 ubiquitin-protein ligase CHFR, EC 6.3.2.-, FLJ10796, FLJ33629, RING finger protein 196, RNF196RNF116
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: AYLIQHPDKSRSEEDVQSMDARNKITQDMLQPKVRRSFSDEEGSSEDLLELSDVDSESSDISQPYVVCRQCPEYRR
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Mitotic Regulators, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 55743
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.