Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chikungunya Virus Antibody (E04C1.02), Novus Biologicals™
SDP

Catalog No. DDX9104P100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX9104P100 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX9104P100 Supplier Novus Biologicals Supplier No. DDX9104P100
Add to Cart
Add to Cart

Human Monoclonal Antibody

Chikungunya virus Monoclonal specifically detects Chikungunya virus in Virus samples. It is validated for Block/Neutralize, ChIP Assay, Immunocytochemistry/Immunofluorescence, Neutralization.

Specifications

Antigen Chikungunya virus
Applications Blocking Assay, ChIP Assay, Immunocytochemistry, Immunofluorescence, Neutralization
Classification Monoclonal
Clone E04C1.02
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence, Neutralization, Chromatin Immunoprecipitation (ChIP), Block/Neutralize
Gene Alias Chikungunya
Host Species Human
Immunogen Chikungunya
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.