Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chikungunya virus Mouse anti-Virus, Clone: 3E7b, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $423.68
Specifications
Antigen | Chikungunya virus |
---|---|
Clone | 3E7b |
Concentration | Please refer to lot specific datasheet. |
Applications | ELISA, Immunocytochemistry, Neutralization, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF2052-25
|
MilliporeSigma
MABF205225UL |
25 μL |
Each for $192.35
|
|
|||||
MABF2052-100
|
MilliporeSigma
MABF2052100UL |
100 μL |
Each for $423.68
|
|
|||||
Description
Anti-Chikungunya virus, clone 3E7b is a mouse monoclonal antibody that detects E2 protein of Chikungunya virus and has been tested for use in ELISA, Immunocytochemistry, Neutralizing asssay, and Western Blotting.Specifications
Chikungunya virus | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Virus | |
IgM κ | |
Purified by ion-exchange chromatography | |
Clone 3E7b detects Chikungunya virus in cells infected with this virus. |
3E7b | |
ELISA, Immunocytochemistry, Neutralization, Western Blot | |
Unconjugated | |
Mouse | |
Inflammation and Immunology | |
Chikungunya virus | |
Primary | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title