Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHIP/STUB1 Rabbit anti-Human, Clone: 6, Novus Biologicals™
SDP

Catalog No. NB021694 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021694 50 μg
NB021693 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021694 Supplier Novus Biologicals Supplier No. NBP29011550UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CHIP/STUB1 Monoclonal specifically detects CHIP/STUB1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CHIP/STUB1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500
Formulation 0.2 um filtered solution in PBS
Gene Alias Antigen NY-CO-7, Carboxy terminus of Hsp70-interacting protein, CHIPSTIP1 homology and U box-containing protein 1, CLL-associated antigen KW-8, E3 ubiquitin-protein ligase CHIP, EC 6.3.2.-, heat shock protein A binding protein 2 (c-terminal), HSPABP2, NY-CO-7, SDCCAG7, serologically defined colon cancer antigen 7, STIP1 homology and U-box containing protein 1, STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase, UBOX1
Host Species Rabbit
Immunogen Recombinant Human CHIP/STUB1 protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10273
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.