Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHIPS Antibody (JNC1) - N-Terminus, Novus Biologicals™
SDP

Catalog No. NB119868 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119868 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119868 Supplier Novus Biologicals Supplier No. NBP311243
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CHIPS Monoclonal specifically detects CHIPS in Bacteria samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunoassay.

Specifications

Antigen CHIPS
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, Immunoassay
Classification Monoclonal
Clone JNC1
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias Chemotaxis Inhibiting Protein of Staphylococcus aureus, Chemotaxis inhibitory protein, Chemotaxis-inhibiting protein, CHP, chs
Host Species Mouse
Immunogen Monoclonal antibody JNC1 reacts with the N-terminus of CHIPS.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3921863
Target Species Bacteria
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.