Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ CHIRALPAK™ HSA HPLC Column, 5 μm Particle Size

Catalog No. 111005869 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
5 cm
10 cm
15 cm
Diameter (Metric):
2 mm
3 mm
4 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-100-5869 10 cm 4 mm
11-100-5849 10 cm 2 mm
11-100-5991 5 cm 3 mm
11-100-5871 10 cm 3 mm
11-100-5990 5 cm 4 mm
11-100-5792 15 cm 4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 11-100-5869 Supplier MilliporeSigma™ Supelco™ Supplier No. 58450AST
Add to Cart
Add to Cart

CHIRALPAK™ HSA, which uses human serum albumin as the chiral selector, is highly selective for acidic racemates, preferably weak and strong acids, zwitterionic and non-protolytic (neutral) compounds.

CHIRALPAK™ HSA, which uses human serum albumin as the chiral selector, is highly selective for acidic racemates, preferably weak and strong acids, zwitterionic and non-protolytic (neutral) compounds. Phosphate buffers (normally 0.01-0.1M, pH 5-7) with addition of organic modifiers are used as mobile phases. Enantioselectivity and retention can be regulated by changing the mobile phase composition. However, the primary use of CHIRALPAK™ HSA is for fast drug/protein binding studies.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4 mm
Length (Metric) 10 cm
Particle Size 5 μm
Stationary Phase Human Serum Albumin
Product Line CHIRALPAK
Matrix Human Serum Albumin Phase
Phase Chiral
Technique Chiral HPLC

CHIRALPAK is a registered trademark of Daicel Corp.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.