Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chitinase 3-like 2 Antibody (2B5), Biotin, Novus Biologicals™
SDP

Catalog No. NB162875 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162875 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162875 Supplier Novus Biologicals Supplier No. NBP260238B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Chitinase 3-like 2 Monoclonal antibody specifically detects Chitinase 3-like 2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen Chitinase 3-like 2
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 2B5
Conjugate Biotin
Formulation PBS
Gene Alias chitinase 3-like 2, chitinase-3-like protein 2, Chondrocyte protein 39, YKL-39YKL39
Host Species Mouse
Immunogen Recombinant human Chitinase 3-like 2 (27-390aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 1117
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.