Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chitinase 3-like 2 Mouse anti-Human, Clone: 2B5, Novus Biologicals™
SDP

Catalog No. NB60238100U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB60238100U 100 μL
NB6023850UL 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB60238100U Supplier Novus Biologicals Supplier No. NBP260238100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chitinase 3-like 2 Monoclonal specifically detects Chitinase 3-like 2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen Chitinase 3-like 2
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 2B5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 2-5 ug, ELISA 1:100 - 1:2000
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias chitinase 3-like 2, chitinase-3-like protein 2, Chondrocyte protein 39, YKL-39YKL39
Gene Symbols CHI3L2
Host Species Mouse
Immunogen Recombinant human CHI3L2 (27-390aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1117
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.