Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chk1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB292262 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292262 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292262 Supplier Novus Biologicals Supplier No. NB100274PCP
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Chk1 Polyclonal specifically detects Chk1 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen Chk1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias Checkpoint, S. pombe, homolog of, 1, CHK1 (checkpoint, S.pombe) homolog, CHK1 checkpoint homolog (S. pombe), CHK1serine/threonine-protein kinase Chk1, EC 2.7.11, EC 2.7.11.1
Host Species Goat
Immunogen The immunogen this antibody was made to, maps to a region between residues 200 and 250 of human Checkpoint Kinase 1 using the numbering given in entry NP_001265.1 (GeneID 1111).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 1111
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.