Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chk2 Antibody (73C175.1.1), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP227403Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227403Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227403Y Supplier Novus Biologicals Supplier No. NBP227403AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chk2 Monoclonal specifically detects Chk2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Chk2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 73C175.1.1
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry-Paraffin
Gene Alias CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, S.pombe) homolog
Gene Symbols CHEK2
Host Species Mouse
Immunogen A portion of amino acids 100-150 of human Chk2 protein was used as immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 11200
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.