Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chk2 Antibody (OTI5C4), Novus Biologicals™
SDP

Catalog No. NBP147695 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147695 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147695 Supplier Novus Biologicals Supplier No. NBP147695
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chk2 Monoclonal specifically detects Chk2 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Chk2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone OTI5C4
Concentration 1.43 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-100, Immunoprecipitation 2 μg/500 μL, Immunohistochemistry-Paraffin 1:150
Gene Alias CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, S.pombe) homolog
Gene Symbols CHEK2
Host Species Mouse
Immunogen Full length human recombinant protein of human CHEK2 (NP_009125) produced in HEK293T cell.
Molecular Weight of Antigen 60.7 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 11200
Test Specificity This antibody is specific for Homo sapiens CHK2 checkpoint homolog (S. pombe) (CHEK2), transcript variant 1.
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.