Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chk2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729118 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9118 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9118 Supplier Proteintech Group Inc Supplier No. 139541AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Chk2 (checkpoint kinase 2) is the mammalian homolog of the Saccharomyces cerevisiae Rad53 and is required for the DNA damage and replication checkpoints. In response to low-dose ionizing radiation (IR), which occurs in an ataxia telangiectasia mutated (ATM)-dependent manner, Chk2 can phosphorylate the mitosis-inducing phosphatase Cdc25C on an inhibitory site, blocking entry into mitosis, and p53 on a regulatory site, causing G1 arrest. In human tissues, Chk2 is homogeneously expressed in renewing cell populations such as epidermis or intestine, heterogeneous in conditionally renewing tissues, and absent or at low level in static tissues such as muscle or brain. Mutations in Chk2 inactivates DNA damage checkpoint pathway involving Chk2 in lung cancer suggesting that reduced expression of Chk2 may be responsible for the development of lung cancer. The Chk2-/- knockout cells are resistant to DNA damage-induced apoptosis, defective for p53 stabilization and for induction of p53-dependent transcripts such as p21 in response to gamma irradiation.

Specifications

Antigen Chk2
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CHEK2
Gene Accession No. O96017, Q9Z265
Gene Alias CDS1, CHEK2, Chk2, HuCds1, LFS2, PP1425, RAD53
Gene Symbols Chek2
Host Species Rabbit
Immunogen Chk2 Fusion Protein Ag5065
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11200, 114212, 50883
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.