Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chk2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Chk2 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9118
![]() ![]() |
Proteintech Group Inc
139541AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Chk2 (checkpoint kinase 2) is the mammalian homolog of the Saccharomyces cerevisiae Rad53 and is required for the DNA damage and replication checkpoints. In response to low-dose ionizing radiation (IR), which occurs in an ataxia telangiectasia mutated (ATM)-dependent manner, Chk2 can phosphorylate the mitosis-inducing phosphatase Cdc25C on an inhibitory site, blocking entry into mitosis, and p53 on a regulatory site, causing G1 arrest. In human tissues, Chk2 is homogeneously expressed in renewing cell populations such as epidermis or intestine, heterogeneous in conditionally renewing tissues, and absent or at low level in static tissues such as muscle or brain. Mutations in Chk2 inactivates DNA damage checkpoint pathway involving Chk2 in lung cancer suggesting that reduced expression of Chk2 may be responsible for the development of lung cancer. The Chk2-/- knockout cells are resistant to DNA damage-induced apoptosis, defective for p53 stabilization and for induction of p53-dependent transcripts such as p21 in response to gamma irradiation.Specifications
Chk2 | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O96017, Q9Z265 | |
11200, 114212, 50883 | |
Chk2 Fusion Protein Ag5065 | |
Primary | |
-20°C | |
CHEK2 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CDS1, CHEK2, Chk2, HuCds1, LFS2, PP1425, RAD53 | |
Chek2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title