Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHL-1/L1CAM-2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB082918 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB082918 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB082918 Supplier Novus Biologicals Supplier No. NBP298946F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHL-1/L1CAM-2 Polyclonal specifically detects CHL-1/L1CAM-2 in Human samples. It is validated for ELISA.

Specifications

Antigen CHL-1/L1CAM-2
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), FLJ44930, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CHL-1/L1CAM-2 (NP_006605.2; Met 1-Gln 1080).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10752
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.