Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia Lps Mouse anti-Bacteria, Clone: Tu-300, Novus Biologicals™
SDP

Catalog No. NB10062450 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062450 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062450 Supplier Novus Biologicals Supplier No. NB10062450

Mouse Monoclonal Antibody

Chlamydia Lps Monoclonal specifically detects Chlamydia Lps in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Chlamydia Lps
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone Tu-300
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence
Host Species Mouse
Immunogen Made to Bacterial CHLAMYDIA LPS
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product is specific for Chlamydia lipopolysaccharide (LPS). LPS is a common feature of the outer envelope of gram negative bacteria, which acts as a potent endotoxin, triggering an innate immune response. Studies have shown however, that whilst the LPS of Chlamydia trachomatis does evoke an immune response, it displays only weak endotoxic activity when compared to that of other bacteria such as Salmonella minnesota or Neisseria gonorrhoeae. This antibody has not been cross-adsorbed and may react with LPS from related species.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.