Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia Momp Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10065054 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065054 0.5 mL
NB017688 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10065054 Supplier Novus Biologicals Supplier No. NB10065054
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Chlamydia Momp Polyclonal specifically detects Chlamydia Momp in Bacteria samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen Chlamydia Momp
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 4.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunohistochemistry-Frozen 1:10-1:500
Formulation Phosphate Buffered Saline with 0.09% Sodium Azide
Gene Alias Major Outer Membrane Protein, MOMP, omp1, omp1F, Omp1L1, ompA, ompIL2
Gene Symbols OMPA
Host Species Goat
Immunogen Purified MOMP (major outer membrane protein) of Chlamydia trachomatis strain L2.
Purification Method IgG purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes L2 MOMP and purified EBs (all serovars, A-K, L1-L3).
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.